missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ CD4

Product Code. 30283706
Change view
Click to view available options
Quantity:
15 mL vial
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283706 15 mL vial 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30283706 Supplier Gibco™ Supplier No. A56994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CTS Detachable Dynabeads CD4 for positive isolation of CD4+ T cells and active release of Dynabeads post-isolation.

Gibco CTS Detachable Dynabeads CD4 are intended for ex vivo isolation of CD4+ T cell subsets. Combined with CTS Detachable Dynabeads Release Buffer (Cat. No. A5588301, A5588302, or A5588303), it is a first-of-its-kind cGMP cell selection technology with an active release mechanism for process development, clinical trial, and commercial manufacturing use. This product is also commonly used with CTS Detachable Dynabeads CD8 (Cat. No. A56995).

Features of CTS Detachable Dynabeads CD4 include:

  • Empower users with process flexibility by allowing active release of beads from target cells at any time
  • Optimize target cell purity and yield thanks to the active release mechanism that gives users more control
  • Provide an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enable consistent performance in light of biological variability within the starting material
  • Suitable for both autologous and allogeneic cell therapy manufacturing processes

These Detachable Dynabeads are intended for use with CTS Detachable Dynabeads Release Buffer, which has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of the beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process.

Based on a recommended 4-to-1 bead-to-target-cell ratio (not total nucleated cell number), one 15 mL vial can theoretically isolate up to 1.5 x 109 CD4+ T cells.

CTS Detachable Dynabeads also include a Drug Master File (DMF) to support cross-referencing in regulatory submissions prior to clinical trial and commercial manufacturing use.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 x 108 beads/mL
Content And Storage CTS™ Detachable Dynabeads™ CD4, 15 mL.

Store at 2°C to 8°C with vial upright to keep beads in suspension.
Color Brown
Material Polystyrene
Product Type Cell Isolation Reagent
For Use With (Equipment) CTS™ DynaCellect™ Magnetic Separation System
Product Line CTS Detachable Dynabeads
Purity or Quality Grade GMP-grade
Quantity 15 mL vial
Shipping Condition Ships at 2°C to 8°C with wet (not frozen) gel packs. Vial must remain upright to keep beads in suspension.
Volume (Metric) 15 mL
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.