missing translation for 'onlineSavingsMsg'
Learn More

CTS-1027, MedChemExpress

Product Code. 16456637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456637 5 mg 5mg
16460277 50 mg 50mg
16453918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456637 Supplier Medchem Express Supplier No. HY10398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CTS-1027 is a potent small molecule inhibitor of MMPs, with IC50s of 0.3 nM, 0.5 nM for MMP2, MMP13, respectively, and has > 1,000 fold selectivity over MMP1.

CTS-1027 is a potent small molecule inhibitor of MMPs, with IC50s of 0.3 nM, 0.5 nM for MMP2, MMP13, respectively, and has > 1,000 fold selectivity over MMP1.

Specifications

Chemical Name or Material CTS-1027
CAS 193022-04-7
Color White
Purity Grade Notes Research
Molecular Formula C19H20ClNO6S
Quantity 5 mg
Synonym Ro 1130830 RS 130830
Solubility Information DMSO : ≥ 100 mg/mL (234.81 mM)
SMILES O=C(NO)C1(CCOCC1)CS(=O)(C2=CC=C(C=C2)OC3=CC=C(C=C3)Cl)=O
Molecular Weight (g/mol) 425.88
Formula Weight 425.88
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.