missing translation for 'onlineSavingsMsg'
Learn More

Origene Technologies CTLA4 Monoclonal Antibody (OTI10C8), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813249S

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17538034

  • 2453.92 NOK / 30µL
Estimated Shipment: 19-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CTLA4 Monoclonal antibody specifically detects CTLA4 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CTLA4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P16410
Mouse
Protein A/G
RUO
1493
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry
OTI10C8
Unconjugated
CTLA4
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
Full length human recombinant protein of human CTLA4 (NP_005205) produced in HEK293T cell.
30 μL
Primary
Human
Antibody
IgG1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers