missing translation for 'onlineSavingsMsg'
Learn More

CTLA4 Monoclonal Antibody (OTI10C8), TrueMAB™, OriGene

Codice prodotto. 17518034
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
30 μL
missing translation for 'unitSize'
100µL
30µL
Codice prodotto. Quantity unitSize
17518034 100 μL 100µL
17538034 30 μL 30µL
2 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 17518034

missing translation for 'mfr': Origene Technologies TA813249

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Questo articolo non è restituibile. Consulta la politica di reso

Mouse Monoclonal Antibody

CTLA4 Monoclonal antibody specifically detects CTLA4 in Human samples. It is validated for Flow Cytometry

Specifica

Antigen CTLA4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI10C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTLA4
Gene Accession No. P16410
Gene Alias ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
Host Species Mouse
Immunogen Full length human recombinant protein of human CTLA4 (NP_005205) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.