Learn More
BD Biosciences™ CS&T Beads (CE/IVD)
Product Code. 15806349
Change view
Click to view available options
No. of Tests:
150 Tests
50 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
15806349 50 Tests Each
15816349 150 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15806349 Supplier BD Biosciences™ Supplier No. 656504
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Provide a standardized method to perform quality control of the instrument’s optics, electronics, and fluidics, and for adjusting fluorescence compensation.
  • Used on a BD flow cytometer.
  • Provides a standardized method to perform quality control of the instrument's optics, electronics, and fluidics, and for adjusting fluorescence compensation.
  • On some BD instruments, BD CS&T beads are also used for adjusting detector voltages.
  • Suspension of fluorospheres with uniform and stable size and fluorescence intensity.
  • Used for instrument quality control (QC) to characterize, track, and report performance measurements of supported flow cytometers.
  • Dyed with fluorochromes that are excited by the cytometer's lasers.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store vials at 2°C–8°C and protect from light.
No. of Tests 50 Tests
Stability The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC
Product Type Fluorescent Polystyrene Beads