Learn More
Asecos™ CS-CLASSIC Two-Door Yellow Safety Storage Cabinet
Product Code. 15546875 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
100 kg Shelves, 60 kg Insert, 33 L Sump, Max Load 600 kg
30 kg/33 L Shelves, Max Load 600 kg
Unit Size:
Each
Product Code. Load Bearing Capacity unitSize
15546875 30 kg/33 L Shelves, Max Load 600 kg Each
15896024 100 kg Shelves, 60 kg Insert, 33 L Sump, Max Load 600 kg Each
2 options
Product Code. 15546875
Brand: Asecos™ 3049504030516
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and proper storage of non-flammable chemicals and toxic substances in working areas.
  • Robust construction and longevity – sturdy all-welded construction for high industrial requirements
  • High-quality, scratch-resistant outer body
  • Double-walled door construction
  • In accordance with TRGS 510
  • Available with double or single wing doors
  • Modern bicolor design
  • Ventilation – ready for connection to a technical exhaust system (DN 75), natural ventilation at the cabinet bottom
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes height-adjustable shelves, pull-out shelves and bottom collecting sumps
  • Powder-coated textured surface
  • No unauthorized use – doors lockable with cylinder locking, integrated into the turning handle
Specifications
Certifications/Compliance TRGS 510
Configuration 4 x 30 kg/33 L powder-coated sheet steel pull-out shelves with PP sump
Dimensions (D x W x H) Interior 493 x 996 x 1835 mm
Dimensions (D x W x H) 520 x 1055 x 1950 mm
Door Style Wing
For Use With (Application) Storage of non-flammable and toxic substances
Load Bearing Capacity 30 kg/33 L Shelves, Max Load 600 kg
No. of Doors 2
No. of Shelves 4
Shelf Type Pull-out shelf
Surface Coating Powder-coated
Weight (Metric) 140.5 kg
Color Anthracite Gray, Warning Yellow
Depth (English) Exterior 20.47 in.
Height (English) Exterior 76.77 in.
Height (Metric) Opening 1835 mm
Material Steel
Product Type Safety Storage Cabinet
Width (English) Exterior 41.54 in.