Learn More
Asecos™ CS-CLASSIC-G One-Door Two-Window Safety Chemical Storage Cabinets
Product Code. 15711008 Shop All asecos Products
Change view
Click to view available options
Color:
Anthracite Gray, Warning Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15711008 Anthracite Gray, Warning Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15711008 Supplier Asecos™ Supplier No. 3056304030579
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For the safe and proper storage of non-flammable chemicals and toxic substances in working areas.
  • Robust construction and longevity – sturdy all-welded construction for high industrial requirements
  • High-quality, scratch-resistant outer body
  • Double-walled door construction
  • In accordance with TRGS 510
  • Available with double or single wing doors with fixed windows
  • Modern bicolor design – body in light gray, wing doors in a variety of colors
  • Ventilation – ready for connection to a technical exhaust system (DN 75), natural ventilation at the cabinet bottom
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • Powder-coated textured surface
  • No unauthorized use – doors lockable with cylinder locking, integrated into the turning handle
Specifications
Configuration 4 x 30 kg/11 L powder-coated sheet steel pull-out shelves with PP sump
Dimensions (D x W x H) Interior 493 x 486 x 1835 mm
Dimensions (D x W x H) 520 x 545 x 1950 mm
Door Style Wing, 2 Windows
For Use With (Application) Storage of non-flammable chemicals and toxic substances
Load Bearing Capacity Pull-out shelves: 30 kg/11 L each, Maximum load: 600 kg
No. of Doors 1
No. of Shelves 4
Shelf Type Pull-out shelf
Surface Coating Powder-coated
Weight (Metric) 105.5 kg
Color Anthracite Gray, Warning Yellow
Depth (English) Exterior 20.47 in.
Height (English) Exterior 76.77 in.
Height (Metric) Opening 1835 mm
Material Steel
Product Type Safety Storage Cabinet
Width (English) Exterior 21.46 in.