Learn More
Witeg™ Crystallizing Dish without Spout
Product Code. 11867860
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11867860 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11867860 Supplier Witeg™ Supplier No. 5 536 100 B
  • ONLINE EXCLUSIVE PRICE
    75.89 NOK
    59.71 NOK / Pack of 1
    Save 16.18 NOK
    21% Off
Estimated Shipment: 24-02-2027
Ideal for storage, crystallization, evaporation and other common laboratory applications.
  • Without spout
  • Manufactured from clear borosilicate glass
  • 70 mm diameter, 40 mm height
  • Provided in packs of 10
Specifications
Bottom Shape Flat
Capacity (Metric) 100 mL
Material Borosilicate Glass
Shape Round
Type Crystallizing Dish
For Use With (Application) Crystallizing, evaporation, storage
Height (Metric) 40 mm
Diameter (Metric) 70 mm