Learn More
Crystal Violet (C.I. 42555), Supelco™
Product Code. 15894070
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894070 25 g 25g
15804080 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894070 Supplier Supelco Supplier No. 1014080025
 Restricted Item
Estimated Shipment: 25-11-2026
Indicator ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 548-62-9
Specifications
Melting Point 189°C to 194°C
MDL Number MFCD00011750
Density 1.19 g/cm3 at 20°C
Synonym Crystal violet (C.I. 42555), Hexamethylenepararosaniline chloride, Gentian violet (C.I. 42555), Hexamethyl-p-rosanilinium, Methyl violet 10 B, Crystal violet (C.I. 42555)
Quantity 25 g
Molecular Weight (g/mol) 407.98
Grade Research