Learn More
Multiroir Controlec™ Crystal Polystyrene Storage Drawers
Product Code. 11768303
Change view
Click to view available options
Capacity (Metric):
1.9 kg
7 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11768303 7 kg Each
11798283 1.9 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 11768303 Supplier Multiroir Controlec™ Supplier No. 11103MU07000C
Estimated Shipment: 10-11-2026
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
View the contents of Multiroir Controlec™ Crystal Polystyrene Storage Drawers clearly due to their construction of crystal clear polystyrene. The drawers also feature a slide assembly, with dividers available to order separately.
  • Total visibility
  • Crystal polypropylene transparent;- like glass
  • Slide assembly
  • Drawer separators available separately
Specifications
Capacity (Metric) 7 kg
Description Handle type: Slider assembly
Color Transparent
Material Polystyrene
Shape Rectangular
Product Type Drawer
Quantity 1 Each
Dimensions (L x W x H) 210x365x125 mm
Length (Metric) 365 mm
Width (Metric) 210 mm
Height (Metric) 125 mm