Learn More
Multiroir™ Crystal Polystyrene Minimax Storage Boxes
Product Code. 11724591
Change view
Click to view available options
Dimensions (L x W x H):
178 x 45 mm (L x H)
228 x 178 x 68 mm
228 x 178 x 72 mm
315 x 230 x 70 mm
315 x 45 mm (L x H)
Unit Size:
Each
Product Code. Dimensions (L x W x H) unitSize
11724591 315 x 230 x 70 mm Each
11734591 228 x 178 x 68 mm Each
11744591 228 x 178 x 72 mm Each
11714591 315 x 45 mm (L x H) Each
11758361 178 x 45 mm (L x H) Each
5 options
Product Code. 11724591
Brand: Multiroir™ 42013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Multiroir™ Crystal Polystyrene Minimax Storage Boxes for your storage and transport needs. The clear, glass-like polystyrene enables you to view your contents clearly from the outside. The boxes are delivered complete with a separator kit for additional organization of contents.
  • Transparent as glass
  • Delivered complete with separator kit
  • Various capacities
Specifications
Color Transparent
Material Polystyrene
Shape Rectangular
Product Type Storage Box
Quantity 12 Case
Dimensions (L x W x H) 315 x 230 x 70 mm
Length (Metric) 315 mm
Height (Metric) 70 mm