Learn More
Thermo Scientific™ Cryoware Permanent Markers and Pens
Product Code. 10772711
Change view
Click to view available options
Color:
Assorted (Red, Blue, Green, Black)
Black
No. per Case:
4
10
Unit Size:
Pack of 10
Pack of 100
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. per Case Unit Size
10772711 Black 4 Pack of 4
11361775 Assorted (Red, Blue, Green, Black) - Pack of 4
11301625 Assorted (Red, Blue, Green, Black) - Pack of 100
10584561 Black 10 Pack of 10
10314551 Assorted (Red, Blue, Green, Black) - Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10772711 Supplier Thermo Scientific™ Supplier No. 63130020
  • ONLINE EXCLUSIVE PRICE
    341.22 NOK
    319.78 NOK / Pack of 4
    Save 21.44 NOK
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
  • Permanent marker set of four
  • Extra-fine point
  • Ink color: Black
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Extra Fine
Product Type Permanent Marking Pen
Color Black
Description Cryoware Permanent Marker
Includes Consists of four extra-fine-point pens
For Use With (Application) Used for marking cardboard and Polycarbonate
No. per Case 4
Quantity 1 Pk.