Learn More
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched
Product Code. 12688036 Shop All Epredia Products
Change view
Click to view available options
Product Type:
10 mm Circular Cryocassette
Annular 25mm Cryocassette
Annular 35mm Cryocassette
Crosshatched 25mm Cryocassette
Crosshatched 40mm Cryocassette
Cryobar Cover
Debris Tray
Glass Anti-Roll Plate
Heat Extractor, Double-weight
Heat Extractor, Standard
Narrow Brush
Narrow Shelf
Tool Shelf
Wide Brush
Wide Shelf
For Use With (Equipment):
FSE/FE
FSE/FE/SME/E
Unit Size:
Each
Pack of 4
Pack of 6
Product Code. Product Type For Use With (Equipment) unitSize
12688036 10 mm Circular Cryocassette FSE/FE/SME/E Pack of 4
12698166 Heat Extractor, Standard FSE/FE Each
12087129 Heat Extractor, Double-weight FSE/FE Pack of 4
12027129 Debris Tray FSE/FE/SME/E Each
12658176 Cryobar Cover FSE/FE/SME/E Each
12648186 Wide Shelf FSE/FE/SME/E Each
12658186 Tool Shelf FSE/FE/SME/E Each
12668186 Narrow Shelf FSE/FE/SME/E Each
12047139 Glass Anti-Roll Plate FSE/FE/SME/E Each
12087119 Wide Brush FSE/FE/SME/E Each
12057119 Narrow Brush FSE/FE/SME/E Each
12688166 Crosshatched 40mm Cryocassette FSE/FE/SME/E Pack of 6
12658036 Annular 25mm Cryocassette FSE/FE/SME/E Pack of 4
12668036 Annular 35mm Cryocassette FSE/FE/SME/E Pack of 4
12698176 Crosshatched 25mm Cryocassette FSE/FE/SME/E Each
15 options
Product Code. 12688036
Brand: Epredia™ 77210153
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Epredia™ Cryotome™ Cryostat Accessories.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
  • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
TRUSTED_SUSTAINABILITY
Specifications
Product Type 10 mm Circular Cryocassette
For Use With (Equipment) FSE/FE/SME/E
Diameter (Metric) 10 mm