Learn More
Thermo Scientific™ Cryopreservation Vials, Canes and Canisters
Product Code. 11874760 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cryo Cane
For Use With (Equipment):
12.5 mm dia. Cryo Tubes
2 Ampules
6 Ampules
9.2 and 10 mL Goblets
Unit Size:
6 ampoules
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
11874760 Cryo Cane 6 Ampules 6 ampoules
11884760 Cryo Cane 2 Ampules Each
15422424 Cryo Cane 9.2 and 10 mL Goblets Each
12750348 Cryo Cane 12.5 mm dia. Cryo Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11874760 Supplier Thermo Scientific™ Supplier No. 4000211
  • ONLINE EXCLUSIVE PRICE
    251.00 NOK
    232.81 NOK / 6 ampoules
    Save 18.19 NOK
    7% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
  • Maximize your cryopreservation storage capacities with a range of vials, canes and canisters.
  • Maximize your cryopreservation storage capacities with a range of vials, canes and canisters.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type Cryo Cane
    Capacity (English) 4.23 cu. ft.
    Capacity (Metric) 1.2 mL
    Color Silver
    For Use With (Equipment) 6 Ampules
    Holds Ampules (6)