Learn More
Thermo Scientific™ CryoPlus™ Canisters, Frames and Dividers
Product Code. 15227476 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
CryoPlus 1
CryoPlus 1, 2, 3, 4
CryoPlus 2
CryoPlus 3
CryoPlus 4
Holds:
Bags (25 mL)
Fenwal 4R5461 Bags (250 mL)
Fenwal 4R5462 Bags (500 mL)
Fenwal 4R9951 Bags (50 mL)
Gambro DF-200 Bags (500 mL)
Gambro DF-700 Bags (700 mL)
Vials (2 mL)
Type:
Canister
Canister Cane Storage, Vapor Phase
Frames, Liquid Phase
Frames, Vapor Phase
Platform Divider
Sliding Canister
Unit Size:
1 set
Each
Pack of 6
Product Code. For Use With (Equipment) Holds Type unitSize
15227476 CryoPlus 1, 2, 3, 4 Fenwal 4R9951 Bags (50 mL) Frames, Vapor Phase Pack of 6
11841200 CryoPlus 3 Gambro DF-200 Bags (500 mL) Platform Divider Each
11851200 CryoPlus 3 Gambro DF-700 Bags (700 mL) Platform Divider Each
11861200 CryoPlus 4 Gambro DF-200 Bags (500 mL) Platform Divider Each
11857993 CryoPlus 2 Fenwal 4R5462 Bags (500 mL) Platform Divider Each
10705725 CryoPlus 1, 2, 3, 4 Gambro DF-200 Bags (500 mL) Canister Each
17843394 CryoPlus 1, 2, 3, 4 Gambro DF-700 Bags (700 mL) Canister Each
10091584 CryoPlus 1, 2, 3, 4 Fenwal 4R5461 Bags (250 mL) Canister Each
10386764 CryoPlus 1, 2, 3, 4 Fenwal 4R5462 Bags (500 mL) Canister Each
15549251 CryoPlus 1, 2, 3, 4 Gambro DF-200 Bags (500 mL) Canister Each
15542066 CryoPlus 1, 2, 3, 4 Gambro DF-700 Bags (700 mL) Canister 1 set
11821200 CryoPlus 1, 2, 3, 4 Gambro DF-200 Bags (500 mL) Frames, Liquid Phase Each
11831200 CryoPlus 1, 2, 3, 4 Fenwal 4R5462 Bags (500 mL) Frames, Liquid Phase Each
15700797 CryoPlus 1, 2, 3, 4 Fenwal 4R5461 Bags (250 mL) Frames, Liquid Phase Each
11847993 CryoPlus 1, 2, 3, 4 Fenwal 4R5462 Bags (500 mL) Frames, Liquid Phase Each
15511701 CryoPlus 1, 2, 3, 4 Vials (2 mL) Canister Cane Storage, Vapor Phase Each
12790348 CryoPlus 1, 2, 3, 4 Fenwal 4R9951 Bags (50 mL) Canister Each
15247476 CryoPlus 1 Fenwal 4R9951 Bags (50 mL) Platform Divider Each
15366524 CryoPlus 1, 2, 3, 4 Fenwal 4R9951 Bags (50 mL) Frames, Liquid Phase Each
15855232 CryoPlus 1, 2, 3, 4 Bags (25 mL) Sliding Canister Each
20 options
Product Code. 15227476
Brand: Thermo Scientific™ 4000660
Only null left
Add to basket
Add to basket
Maximize capacity and protect sample integrity with Thermo Scientific™ CryoPlus™ Canisters, Frames and Dividers, for use with CryoPlus™ LN2 cryopreservation storage systems.
Maximize capacity and protect sample integrity. Thermo Scientific™ CryoPlus™ Canisters, Frames and Dividers are for use with CryoPlus LN2 cryopreservation storage systems. Ideal for storing blood, blood components and bone marrow.
Frames (liquid)
  • 50mL bag capacity, Fenwal 4R9951, holds 7 canisters
  • 250mL bag capacity, Fenwal 4R5461, holds 4 canisters
  • 500mL bag capacity, Fenwal 4R5462, holds 4 canisters
  • 500mL bag capacity, Gambro DF-200, holds 4 canisters
  • 700mL bag capacity, Gambro DF-700, holds 4 canisters
Canisters
  • 50mL bag capacity, Fenwal 4R9951, swing arm canister
  • 250mL bag capacity, Fenwal 4R5461, sliding canister
  • 500mL bag capacity, Fenwal 4R5462, sliding canister
  • 500mL bag capacity, Gambro DF-200, sliding canister
  • 500mL bag capacity, Gambro DF-200, swing arm canister
  • 700mL bag capacity, Gambro DF-700, sliding arm canister
  • 700mL bag capacity, Gambro DF-700, swing arm canister
Platform Dividers
  • 50mL Fenwal 4R9951 for frames and canisters
  • 250mL Fenwal 4R5461 for frames and canisters
  • 500mL Fenwal 4R5462 for frames and canisters
  • 500mL Gambro DF-200 for frames and canisters
  • 700mL Gambro DF-700 for frames and canisters
TRUSTED_SUSTAINABILITY
Specifications
Type Frames, Vapor Phase
Capacity (Metric) 700 mL
Door Style Sliding
For Use With (Equipment) CryoPlus 1, 2, 3, 4
Holds Fenwal 4R9951 Bags (50 mL)