Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
Product Code. 30968820 Shop All Thermo Scientific Products
Click to view available options
Type:
Delmed 2030-2
Gambro DF-200
Gambro DF-700
Nexell R4R9955 (Fenwal 4R5462)
Holds:
Bags (2)
Bags (4)
Unit Size:
Each
Product Code. 30968820
Brand: Thermo Scientific™ 4000317
Perform controlled-rate freezing applications with a controlled-rate freezer and these CryoMed bag freezing presses.
Perform controlled-rate freezing applications with a controlled-rate freezer and these Thermo Scientific™ CryoMed™ Bag Freezing Presses.
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2 to 4 bags.
TRUSTED_SUSTAINABILITY
Specifications
Type Delmed 2030-2
For Use With (Equipment) CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Holds Bags (4)