Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
Shop All Thermo Scientific Products
Click to view available options
Type:
Delmed 2030-2
Gambro DF-200
Gambro DF-700
Nexell R4R9955 (Fenwal 4R5462)
Holds:
Bags (2)
Bags (4)
Unit Size:
Each
Description
Perform controlled-rate freezing applications with a controlled-rate freezer and these Thermo Scientific™ CryoMed™ Bag Freezing Presses.
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2 to 4 bags.
Specifications
Specifications
| Description | Hält 2 Beutel |
| Type | Gambro DF-700 |
| For Use With (Equipment) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 |
| Holds | Bags (2) |