Learn More
R&D Systems™ CryoDefend-Stem Cells (5 x 10 mL)
Product Code. 18491540
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18491540 5 x 10 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18491540 Supplier R&D Systems™ Supplier No. CCM018
  • ONLINE EXCLUSIVE PRICE
    3115.00 NOK
    2944.20 NOK / 1 sets
    Save 170.80 NOK
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Media for defined, protein-free cryopreservation of stem cells
Cryopreservation is essential for the banking of valuable stem cell cultures. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
  • Reduce experimental variability by using fully defined media
  • Supports post-thaw cell viability better than conventional freezing media
  • Optimized for cryopreservation of stem cells
  • Suitable for all stem cell types
TRUSTED_SUSTAINABILITY
Specifications
Product Type CryoDefend-Stem Cells
Product Line R&D Systems
Quantity 5 x 10 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.