Learn More
Eppendorf CryoCube F740hiw
GREENER_CHOICE
Product Code. 17803651
Click to view available options
Capacity (Metric):
740 L
Unit Size:
Each
Product Code. 17803651
Brand: Eppendorf F740320141
The Eppendorf CryoCube F740 series supports secure sample protection and minimized running costs. With up to 576 boxes, the CryoCube F740hiw freezer is the water-cooled version of the 740 L class. The PhysioCare concept provides easy and comfortable usage in respect to ergonomics.
  • Touchscreen interface
  • Consumes only 11.7 kWh/day (green)
  • Holds up to 57.600 samples
  • Pull-down time to -80°C: 4 h 10 min.
  • Interior: Stainless steel; exterior: powder coated steel
  • Polyurethane foam + Vacuum insulation panels (body + door)
  • 3 compartments with 3 insulated and sealed inner doors
  • Left-handed outer door, water-cooled
  • Compatible with CO2/LN2 injection system (Type 'narrow') as backup for highest sample safety
TRUSTED_SUSTAINABILITY
Specifications
Description CryoCube F740hiw, ULT freezer, 740L, VIP, WC, DoRi, 5, 230V/50Hz (EU)
Type Freezers
Capacity (English) 26.1 cu. ft.
Capacity (Metric) 740 L
Temperature Resolution 1°C
Door Style Hinged
No. of Doors 1
Inner Doors 3
Hinge Side Left
Cabinet Material Steel
Chamber Material Stainless Steel
Insulation Vacuum, Polyurethane Foam
Shelves 5
Style Freestanding
Display Touchscreen interface
Casters Yes
Water Cooled Yes
Dimensions (D x W x H) Interior 62.1 × 86.5 × 139 cm / 24.5 × 34.1 × 54.7 in.
Depth (English) Exterior 38.6 in.
Depth (English) Interior 24.5 in.
Depth (Metric) Exterior 98.0 cm
Depth (Metric) Interior 62.1 cm
Height (English) Exterior 77.7 in.
Height (English) Interior 54.7 in.
Height (Metric) Exterior 197.3 cm
Height (Metric) Interior 139 cm
Width (English) Exterior 43.7 in.
Width (English) Interior 34.1 in.
Width (Metric) Exterior 111 cm
Width (Metric) Interior 86.5 cm
Monitoring Options Touch Interface
Access Security Key Lock
Backup Systems CO2 or LN2 Optional
Amperage 0.25
Plug Type EU
Voltage 220 V
Electrical Requirements 230V/50Hz (EU)
Compressor 2-stage
Refrigerant R290 / R170
Holds 57,600 Samples
Rack Capacity 18