Learn More
Thermo Scientific™ Cryo Tong
Product Code. 12780348 Shop All Thermo Scientific Products
Change view
Click to view available options
Weight (English):
1 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
12780348 1 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12780348 Supplier Thermo Scientific™ Supplier No. 4000388
  • ONLINE EXCLUSIVE PRICE
    2555.00 NOK
    2445.04 NOK / Pack of 1
    Save 109.96 NOK
    4% Off
 Restricted Item
In Stock
Safely retrieve cryopreservation storage products using the Thermo Scientific™ Cryo Tong.
 Shipping dimensions: 12 x 7 x 6 in. box.
TRUSTED_SUSTAINABILITY
Specifications
Description Cryo Tong
Type Cryo Tong
For Use With (Equipment) All CryoMed Controlled-Rate Freezers
Weight (English) 1 lb.