Learn More
Spex CertiPrep™ Cryo-Stations
Product Code. 12567326 Shop All SPEX Certiprep Products
Change view
Click to view available options
For Use With (Equipment):
Cryo-Blocks
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12567326 Cryo-Blocks Each
1 options
Product Code. 12567326
Brand: Spex CertiPrep™ 2600
This item requires a price update prior to being ordered.
Request a Price for this item here.
Keep samples cold during sample manipulation and prior to or after grinding
A dedicated holding station for temperature-sensitive samples that must be kept chilled during processing with SPEX SamplePrep homogenisers. Closed system is filled with liquid nitrogen automatically or manually. Either sample vials and cryoblocks can be placed in the Cryostation to keep samples cold.
  • Holds two titre plates or three cryoblocks
  • Keep samples cold during sample manipulation and prior to or after grinding
Specifications
Product Type Cryo-Station
For Use With (Equipment) Cryo-Blocks
Includes Cutting board for preparing temperature sensitive samples
Model 2600