Learn More
Welch™ CRVpro™ Direct Drive Rotary Vane Vacuum Pump
Product Code. 16238250 Shop All Welch Ilmvac Products
Change view
Click to view available options
Model:
CRVpro 16
CRVpro 24
CRVpro 30
CRVpro 4
CRVpro 6
CRVpro 8
Air Displacement:
10.7 CFM, 305 L/min.
15.3 CFM, 436 L/min.
18.4 CFM, 551 L/min.
2.8 CFM, 78 L/min.
3.5 CFM, 100 L/min.
4.7 CFM, 133 L/min.
Dimensions (L x W x H):
18.2 x 6.1 x 9.1 in.
22.4 x 8.1 x 11.4 in.
Weight (English):
46.3 lb.
48.5 lb.
49.6 lb.
81.6 lb.
84.9 lb.
87 lb.
Unit Size:
Each
Product Code. Model Air Displacement Dimensions (L x W x H) Weight (English) unitSize
16238250 CRVpro 16 10.7 CFM, 305 L/min. 22.4 x 8.1 x 11.4 in. 81.6 lb. Each
16228250 CRVpro 4 2.8 CFM, 78 L/min. 18.2 x 6.1 x 9.1 in. 46.3 lb. Each
15706275 CRVpro 6 3.5 CFM, 100 L/min. 18.2 x 6.1 x 9.1 in. 48.5 lb. Each
15852782 CRVpro 8 4.7 CFM, 133 L/min. 18.2 x 6.1 x 9.1 in. 49.6 lb. Each
16248250 CRVpro 24 15.3 CFM, 436 L/min. 22.4 x 8.1 x 11.4 in. 84.9 lb. Each
16258250 CRVpro 30 18.4 CFM, 551 L/min. 22.4 x 8.1 x 11.4 in. 87 lb. Each
6 options
Product Code. 16238250
Brand: Welch™ 316200
Only null left
Add to basket
Add to basket
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
  • Welch Gardner Denver CRVpro Two-Stage Rotary Vane Vacuum Pump
  • The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
  • The pump impresses with its long service life, absolute reliability and its low maintenance requirements.
  • This has been enabled thanks to 3 major innovations:
    • Its low running temperature, which slows down potential corrosion processes and reduces the oil consumption.
    • Its large oil reservoir ensures a low chemical concentration, which contributes to the prolongation of the oil exchange intervals.
    • Its special inside coating increases the chemical resistance of the pump.
  • Scope of delivery: Pump with motor, 3 line cords (EU/UK/US), 2 liters of oil, centering ring with dirt trap, centering ring, 2 clamping rings.
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 0.005 torr
Horsepower 1 hp
Includes Pump with Motor, 3 Line Cords (EU/UK/US), 2 L of Oil, Centering Ring with Dirt Trap, Centering Ring, 2 Clamping Rings
Motor Speed 1450 rpm
Certifications/Compliance cUL and CE
Dimensions (L x W x H) 22.4 x 8.1 x 11.4 in.
Model CRVpro 16
Weight (English) 81.6 lb.
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Frequency 50/60 Hz
Pump Type Rotary Vane Vacuum Pump
Air Displacement 10.7 CFM, 305 L/min.
Oil Capacity 2400 mL
Maximum Pressure 3 x 10-3 mbar (2 x 10-3 torr)
Voltage 230 V