Learn More
Nickel Electro™ Nickel Crucibles without Lids
Shop All Nickel-Electro Products
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Each
Description
- Laboratory nickel crucible
- Nickel 201 spec (minimum 99.6% nickel content)
- Formed to shape from one nickel billet
- Bright finish inside and out
- Supplied in standard packs of 10
Specifications
Specifications
| Material | Nickel |
| Capacity (Metric) | 10 mL |
| Diameter (Metric) Outer | 25 mm |
| Height (Metric) | 30 mm |