Learn More
Haldenwanger CRUCIBLE ALSINT ALumINA TALL FORM CAPACITY 500ml
Brand:  Haldenwanger L ALSINTTIEGEL*C/8
CRUCIBLE ALSINT ALumINA TALL FORM CAPACITY 500ml
Product Code. 10018560
  • ONLINE EXCLUSIVE PRICE
    4950.00 NOK
    4586.55 NOK / Each
    Save 363.45 NOK
    7% Off
Only null left
Estimated Shipment: 27-01-2026