Learn More
Crown ether/18-Crown-6, Sigma-Aldrich™
Product Code. 15836500
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15836500 5 g 5g
15809730 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15836500 Supplier Sigma Aldrich Supplier No. 8116840005
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17455-13-9
Specifications
Melting Point 36°C to 39°C
MDL Number MFCD00005113
Quantity 5 g
Synonym Crown ether/18-Crown-6, 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6, Crown ether/18-Crown-6
Molecular Weight (g/mol) 264.32
Grade Research