missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRLS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51055-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein is still unknown. The gene of human hypothetical protein (LOC54675) is up-regulated in colon cancer metastasis.Specifications
| CRLS1 | |
| Polyclonal | |
| Unconjugated | |
| CRLS1 | |
| C20orf155, Cardiolipin synthase, cardiolipin synthase 1, CLS, CLS1, CRLS1, GCD10, Protein GCD10 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54675, 66586 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80ZM8, Q9UJA2 | |
| CRLS1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction