missing translation for 'onlineSavingsMsg'
Learn More

Proteintech CRIPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11211-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898133

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.
Specifications

Specifications

CRIPT
Polyclonal
Unconjugated
CRIPT
CRIPT, HSPC139
Rabbit
Antigen Affinity Chromatography
RUO
56724, 56725, 9419
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70333, Q792Q4, Q9P021
Cript
CRIPT Fusion Protein Ag1708
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers