Learn More
Fisherbrand™ Crimping Heads for Programmable Electronic High Power Crimp Station
Product Code. 15542310 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Cap:
8 mm
11 mm
13 mm
20 mm
Unit Size:
Each
Product Code. Diameter (Metric) Cap unitSize
15542310 11 mm Each
15532340 20 mm Each
15542300 8 mm Each
15562330 13 mm Each
4 options
Product Code. 15542310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Programmable Electronic High Power Crimp Station
  • Crimping Head for Aluminum Caps
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Crimping Head
Diameter (Metric) Cap 11 mm
Material Plastic, Steel
Autoclavable Not Autoclavable
Product Type Crimping Head