Learn More
CRH/CRF Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10944-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The CRH/CRF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRH/CRF. This antibody recognizes human, mouse, rat antigen. The CRH/CRF antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CRH/CRF | |
Polyclonal | |
Unconjugated | |
20 μL | |
1392 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
400 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |