missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CREBL1 (Human) Recombinant Protein

Product Code. 16123491
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16123491

Brand: Abnova™ H00001388P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CREBL1 full-length ORF ( AAH08394.1, 1 a.a. - 318 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MAELMLLSEIADPTRFFTDNLLSPEDWGLQNSTLYSGLDEVAEEQTQLFRCPEQDVPFDGSSLDVGMDVSPSEPPWELLPIFPDLQVKSEPSSPCSSSSLSSESSRLSTEPSSEALGVGEVLHVKTESLAPPLCLLGDDPTSSFETVQINVIPTSDDSSDVQTKIEPVSPCSSVNSEASLLSADSSSQAFIGEEVLEVKTESLSPSGCLLWDVPAPSLGAVQISMGPSLDGSSGKALPTRKPPLQPKPVVLTTVPMPSRAVSPSTTVLLQSLVQPPPGTEEGEKGRAWWLTPVIPALWEAEAGESPEVRSLRPAWPTW

Specifications

Accession Number AAH08394.1
Gene ID (Entrez) 1388
Name activating transcription factor 6 beta
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CREB-RP, CREBL1, FLJ10066, G13
Gene Symbol ATF6B
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.