missing translation for 'onlineSavingsMsg'
Learn More

CREB3L1/OASIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11235-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848173

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. Specifically involved in ER-stress response in astrocytes in the central nervous system (By similarity). May play a role in gliosis. In vitro, binds to box-B element, cAMP response element (CRE) and CRE-like sequences, and activates transcription through box-B element but not through CRE.
Specifications

Specifications

CREB3L1/OASIS
Polyclonal
Unconjugated
CREB3L1
CREB3L1, OASIS
Rabbit
Antigen Affinity Chromatography
RUO
26427, 362165, 90993
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96BA8, Q9Z125
Creb3l1
CREB3L1,OASIS Fusion Protein Ag1757
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers