Learn More
CREB3L1/OASIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11235-2-AP-20UL
 View more versions of this product
Product Code. 16858173
  • 1710.33 NOK / 20µL
Description
Transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. Specifically involved in ER-stress response in astrocytes in the central nervous system (By similarity). May play a role in gliosis. In vitro, binds to box-B element, cAMP response element (CRE) and CRE-like sequences, and activates transcription through box-B element but not through CRE.
Specifications
Specifications
CREB3L1/OASIS
Polyclonal
Unconjugated
CREB3L1
CREB3L1, OASIS
Rabbit
Antigen Affinity Chromatography
RUO
26427, 362165, 90993
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96BA8, Q9Z125
Creb3l1
CREB3L1,OASIS Fusion Protein Ag1757
20 μL
Primary
Mouse, Human, Rat