Learn More
CREB1 Mouse anti-Human, Unconjugated, Clone: 1E11C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67927-1-IG-20UL
 View more versions of this product
Product Code. 17581436
  • 1710.33 NOK / 20µL
Description
The CREB1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CREB1. This antibody recognizes human, mouse, rat antigen. The CREB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CREB1
Monoclonal
Unconjugated
20 μL
1385
IgG1
Western Blot, Immunohistochemistry, ELISA
1E11C1
Mouse
Primary
Human, Mouse, Rat