missing translation for 'onlineSavingsMsg'
Learn More

CREB1 Mouse anti-Human, Unconjugated, Clone: 1E11C1, Proteintech

Product Code. 17571436 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17571436 150 μL 150µL
17581436 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 17571436

Brand: Proteintech 679271IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CREB1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CREB1. This antibody recognizes human, mouse, rat antigen. The CREB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen CREB1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 1E11C1
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1385
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.