Learn More
Proteintech CREB1 Mouse anti-Human, Unconjugated, Clone: 1E11C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67927-1-IG-20UL
Additional Details : Weight : 0.00020kg
Description
The CREB1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CREB1. This antibody recognizes human, mouse, rat antigen. The CREB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CREB1 | |
Monoclonal | |
Unconjugated | |
20 μL | |
1385 | |
IgG1 |
Western Blot, Immunohistochemistry, ELISA | |
1E11C1 | |
Mouse | |
Primary | |
Human, Mouse, Rat |