missing translation for 'onlineSavingsMsg'
Learn More

Creatinine Amidohydrolase from Pseudomonas sp., MP Biomedical

Product Code. 11448771 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 U
Unit Size:
25 units
This item is not returnable. View return policy

Product Code. 11448771

Brand: MP Biomedicals 0215349125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Creatinine Amidohydrolase from Pseudomonas sp. is useful for enzymatic determination of creatinine when coupled with creatine amidinohydrolase, sarcosine dehydrogenase or sarcosine oxidase and formaldehyde dehydrogenase in clinical analysis.

  • Creatininase from Pseudomonas sp. is a homohexameric enzyme with a molecular mass of 28.4 kDa per subunit
  • It is a cyclic amidohydrolase catalysing the reversible conversion of creatinine to creatine
  • Each monomer contains a binuclear zinc centre near the C termini of the β-strands and the N termini of the main α-helices
  • These zinc ions indicate the location of the active site
  • One unit causes the formation of one μmole of creatinine-picrate per minute at pH 7.5 at 37°C
    150 units/mg solid
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 6.5 to 7.5 (Optimum); pH Stability: pH 7.5 to 9.0 (5°C, 16hr).
Packaging Glass
Quantity 25 U
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.