missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine phosphate, disodium salt, 99.12%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 98-100% |
| Formula Weight | 327.2 |
| Chemical Name or Material | Creatine phosphate, disodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction