missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRCP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14348-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CRCP | |
| Polyclonal | |
| Unconjugated | |
| CRCP | |
| CGRP RCP, CGRP receptor component, CGRPRCP, CRCP, HsC17, RCP, RCP9, RNA polymerase III subunit C9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114205, 12909, 27297 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35427, O75575, Q8VHM6 | |
| CRCP | |
| CRCP Fusion Protein Ag5689 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction