Description
The CRB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CRB1. This antibody recognizes mouse antigen. The CRB1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
| Antigen | CRB1 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 23418 |
| Target Species | Mouse |
| Isotype | IgG |