Learn More
Abeba™ Crawler ALU 34555 Shoes
Product Code. 15573744
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15573744 36 1 pair
15523754 37 1 pair
15543754 38 1 pair
15563754 39 1 pair
15573754 40 1 pair
15533764 41 1 pair
15553764 42 1 pair
15563764 43 1 pair
15553824 44 1 pair
15563824 45 1 pair
15573824 46 1 pair
15553844 47 1 pair
15573844 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15573744 Supplier Abeba™ Supplier No. 34555/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3579)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Scratch-resistant coating in toe area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 34555
Toe Material Aluminum