missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Craft Blades

Finger-friendly™ edge cuts materials effectively but is safer to touch than traditional blades

Brand:  Slice™ 10520

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 16453619

  • 301.00 NOK / Pack of 4

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

  • 4 single-edge blades per pack
  • Compatible w/ Slice™ craft handles, scalpel, Precision Knife
  • Compatible tool: Slice™ 10548
  • Finger-friendly™ edge stays sharp 11x longer
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic
  • Safe up to 1600 degrees Celsius
  • Chemically inert, never rusts
  • No-tool blade change
  • Fewer blade changes = fewer injuries
  • Specifications

    Specifications

    White
    Curved Edge, Rounded Tip
    4 single-edge blades per pack
    1.29 in
    0.25 in.
    0.02 oz.
    Zirconium Oxide
    Safety Cutting
    4 Pk.
    32.8 mm
    6.30 mm
    0.55 g
    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers