missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12588-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.Specifications
| CRABP1 | |
| Polyclonal | |
| Unconjugated | |
| CRABP1 | |
| CRABP, CRABP I, CRABP1, CRABPI, RBP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12903, 1381, 25061 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29762, P62965, P62966 | |
| CRABP1 | |
| CRABP1 Fusion Protein Ag3291 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction