missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Crêped Technical Filter Papers

Crêped Technical Filter Papers are mostly used for the rapid filtration of relatively coarse precipitates; because of their crêped structure they provide a larger filtration area than smooth filter papers.

Brand:  Sartorius FT-210375-390390

Additional Details : Weight : 29.83500kg

 View more versions of this product

Product Code. 17304430

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Crêped Filter Papers for Qualitative and Technical Analyses
  • Made of refined pulp and cotton linters with an 95% α-cellulose content
  • Ash content between 0.1 to 0.15%
  • Wet-strengthened
Specifications

Specifications

Pressure or vacuum filtration
Cellulose
Creped Filter Paper
Cotton Linters, Refined Pulp
376
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers