missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Crêped Technical Filter Papers

Crêped Technical Filter Papers are mostly used for the rapid filtration of relatively coarse precipitates; because of their crêped structure they provide a larger filtration area than smooth filter papers.

Brand:  Sartorius FT-3-335-220Q

Additional Details : Weight : 1.43000kg

 View more versions of this product

Product Code. 15992920

  • 870.00 NOK / Pack of 500
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • Crêped Filter Papers for Qualitative and Technical Analyses
  • Made of refined pulp and cotton linters with an 95% α-cellulose content
  • Ash content between 0.1 to 0.15%
  • Wet-strengthened
Specifications

Specifications

Pressure or vacuum filtration
Cellulose
Creped Filter Paper
Cotton Linters, Refined Pulp
603/N
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers