missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26555-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.Specifications
CPT2 | |
Polyclonal | |
Unconjugated | |
CPT2 | |
CPT II, CPT1, CPT2, CPTASE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12896, 1376, 25413 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18886, P23786, P52825 | |
Cpt2 | |
CPT2 Fusion Protein Ag24897 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |