missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26555-1-AP-20UL
This item is not returnable.
View return policy
Description
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.Specifications
| CPT2 | |
| Polyclonal | |
| Unconjugated | |
| CPT2 | |
| CPT II, CPT1, CPT2, CPTASE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12896, 1376, 25413 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18886, P23786, P52825 | |
| Cpt2 | |
| CPT2 Fusion Protein Ag24897 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction