missing translation for 'onlineSavingsMsg'
Learn More

CPOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12211-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849783

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).
Specifications

Specifications

CPOX
Polyclonal
Unconjugated
CPOX
Coprogen oxidase, Coproporphyrinogen oxidase, Coproporphyrinogenase, COX, CPO, CPOX, CPX, HCP
Rabbit
Antigen Affinity Chromatography
RUO
12892, 1371, 304024
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36551, P36552, Q3B7D0
CPOX
CPOX Fusion Protein Ag2858
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers