missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPNE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10126-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11. 21. Alternate splicing results in multiple transcript variants encoding different proteins.Specifications
CPNE1 | |
Polyclonal | |
Unconjugated | |
CPNE1 | |
Copine 1, copine I, COPN1, CPN1, CPNE1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8904 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99829 | |
CPNE1 | |
CPNE1 Fusion Protein Ag0139 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |