Learn More
Chemsplash CPE OverShoe Compressed Polyethylene -Machine Made
Description
Elasticated Overshoe
- 3 GSM Compressed Polyethylene Material
- 16 in. Long
- Elasticated Ankles
- Heat Sealed Seams
- PFAS Free
Description: Elasticated Overshoe in a 3 gram blue compressed polyethylene material. Creates a barrier over your footwear. The 16 in. length offers suitability for all footwear sizing. Ideal for keeping clean footwear clean or keeping clean floors clean from dirty footwear.
Specifications
Specifications
| Size | 3 g |
| Material | Compressed Polyethylene |
| Latex Free | Yes |
| Coating | Polyethylene |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Plain |
| Resistance | Liquid |
| Show More |