Learn More
Cole-Parmer™ CPC Quick-Disconnect Valved NPT(M) Pipe Adapter Insert
Quick Disconnect couplings feature a thumb latch for easy assembly.
Brand: Cole-Parmer™ 06361-66
Additional Details : Weight : 0.01000kg
Description
- CPC Quick-Disconnect NPT(M) Pipe Adapter Insert
- Standard flow quick-disconnect couplings require a body and an insert of the same flow size
- Plug insert into body to connect fitting
- Fitting halves lock into place immediately
- To disconnect fitting halves, simply press the thumb latch on the coupling body
Specifications
Valved NPT(M) Pipe Adapter Insert | |
Autoclavable | |
3/8 in. | |
3/8 in. NPT(M) | |
Buna N | |
28 in. Hg | |
82°C |
Quick-Disconnect x NPT(M) Connection | |
Acetal | |
1/4 in. | |
Autoclave or Ethylene Oxide | |
120 psi | |
180°F |